Gene Transcripts

Gene Symbol
WWC2
Entrez Gene ID
80014
Gene Name
WW and C2 domain containing 2
Gene Aliases
BOMB
Chromosome Location
Chr. 4: 183098677 - 183320777 on Build GRCh38
UniGene ID
Hs.333179
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024949.5 NP_079225.5 11 1590
XM_011532269.2 XP_011530571.1 11 2223
GenBank mRNA AK126057.1 - 10 1255
AK127061.1 - 4 502
BC053873.1 - 2 166
BC107683.1 - 2 187
BX647378.1 - 9 1181

View Full Product Details