Gene Transcripts
- Gene Symbol
- PRR7
- Entrez Gene ID
- 80758
- Gene Name
- proline rich 7 (synaptic)
- Gene Aliases
- Chromosome Location
- Chr. 5: 177445936 - 177456701 on Build GRCh38
- UniGene ID
- Hs.534492
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001174101.1 |
NP_001167572.1
|
2, 3 |
840 |
|
NM_001174102.1 |
NP_001167573.1
|
2, 3 |
757 |
|
NM_030567.4 |
NP_085044.2
|
3, 4 |
925 |
|
XM_011534662.1 |
XP_011532964.1
|
2, 3 |
6663 |
|
XM_011534663.2 |
XP_011532965.1
|
2, 3 |
1401 |
|
XM_017009896.1 |
XP_016865385.1
|
1, 2 |
7451 |
|
XM_017009897.1 |
XP_016865386.1
|
2, 3 |
1633 |
|
GenBank mRNA |
BC004261.1 |
- |
3, 4 |
757 |
|
BC021240.1 |
- |
2, 3 |
748 |
3603954 |
BC024233.2 |
- |
3, 4 |
929 |
4697841 |
BC033649.1 |
- |
1, 2 |
515 |
5538503 |
View Full Product Details