Gene Transcripts

Gene Symbol
ARMCX4
Entrez Gene ID
100131755
Gene Name
armadillo repeat containing, X-linked 4
Gene Aliases
CXorf35, GASP4
Chromosome Location
Chr. X: 101418263 - 101535988 on Build GRCh38
UniGene ID
Hs.729062
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_028407.2 11 2809
NR_045861.1 8 2700
NR_045862.1 7 1834
NR_045863.1 7 1828
NR_045864.1 8 2742
GenBank mRNA AK125041.1 - 7 1632
AK126987.1 - 8 2563
BC032236.1 - 8 2479
BC036206.1 - 7 1830
CR933662.1 - 11 2784

View Full Product Details