Gene Transcripts

Gene Symbol
NKAIN3
Entrez Gene ID
286183
Gene Name
Na+/K+ transporting ATPase interacting 3
Gene Aliases
FAM77D
Chromosome Location
Chr. 8: 62248942 - 62999652 on Build GRCh38
UniGene ID
Hs.654662
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_130764.1 7 1771
XM_017013359.1 XP_016868848.1 6 869
GenBank mRNA AK096949.1 - 7 1771

View Full Product Details