Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_052886.2 | NP_443118.1 | 4 | 2512 | |
XM_011516807.2 | XP_011515109.1 | 3 | 2296 | ||
GenBank mRNA | AK172820.1 | - | 4 | 2421 | |
AL117612.1 | - | 1 | 1803 | ||
AY007723.1 | - | 4 | 2489 |