Gene Transcripts

Gene Symbol
C10orf25
Entrez Gene ID
220979
Gene Name
chromosome 10 open reading frame 25
Gene Aliases
Chromosome Location
Chr. 10: 44993932 - 45000910 on Build GRCh38
UniGene ID
Hs.707791
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039380.3 NP_001034469.2 2 1008
GenBank mRNA AK055129.1 - 2 986

View Full Product Details