Gene Transcripts

Gene Symbol
MAMDC2
Entrez Gene ID
256691
Gene Name
MAM domain containing 2
Gene Aliases
Chromosome Location
Chr. 9: 70043519 - 70226972 on Build GRCh38
UniGene ID
Hs.547172
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153267.4 NP_694999.3 9 1994
NR_125850.1 9 1994
XM_011518471.1 XP_011516773.1 9 2056
XM_011518475.2 XP_011516777.1 9 2056
XM_017014587.1 XP_016870076.1 9 2056
GenBank mRNA AK304040.1 - 9 1754
AK304531.1 - 9 1582
BC015417.2 - 1 58
BC016383.1 - 9 1712 4696722
BC063634.1 - 9 1971 4399866

View Full Product Details