Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_022916.4 | NP_075067.2 | 4 | 535 | |
GenBank mRNA | AF439857.1 | - | 4 | 446 | |
AK315545.1 | - | 4 | 473 | ||
BC016617.1 | - | 4 | 443 | 3855313 | |
BI551252.1 | - | 4 | 488 | ||
BP199163.1 | - | 4 | 535 |