Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_022916.4 | NP_075067.2 | 9, 10 | 1285 | |
GenBank mRNA | AF439857.1 | - | 9, 10 | 1196 | |
AK026048.1 | - | 3, 4 | 406 | ||
AK026840.1 | - | 9, 10 | 1195 | ||
AK315545.1 | - | 9, 10 | 1223 | ||
BC016617.1 | - | 9, 10 | 1193 | 3855313 |