Gene Transcripts

Gene Symbol
ZSCAN5A
Entrez Gene ID
79149
Gene Name
zinc finger and SCAN domain containing 5A
Gene Aliases
ZNF495, ZSCAN5
Chromosome Location
Chr. 19: 56221303 - 56368383 on Build GRCh38
UniGene ID
Hs.177688
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001322061.1 NP_001308990.1 6 1173
NM_001322062.1 NP_001308991.1 5 941
NM_001322064.1 NP_001308993.1 5 1110
NM_001322065.1 NP_001308994.1 5 985
NM_001322066.1 NP_001308995.1 5 1180
NM_001322067.1 NP_001308996.1 5 884
NM_001322068.1 NP_001308997.1 5 1009
NM_001322069.1 NP_001308998.1 5 1180
NM_001322070.1 NP_001308999.1 5 1055
NM_001322072.1 NP_001309001.1 6 1332
NM_001322073.1 NP_001309002.1 5 985
NM_001322074.1 NP_001309003.1 5 1112
NM_001322075.1 NP_001309004.1 5 1180
NM_001322076.1 NP_001309005.1 5 1055
NM_001322077.1 NP_001309006.1 4 599
NM_001322078.1 NP_001309007.1 4 560
NM_024303.2 NP_077279.1 5 1066
XM_005259254.1 XP_005259311.1 5 941
XM_006723377.3 XP_006723440.1 4 1225
XM_011527308.1 XP_011525610.1 4 750
XM_017027298.1 XP_016882787.1 4 1210
XM_017027299.1 XP_016882788.1 4 1307
GenBank mRNA AK098700.1 - 4 698
AK223485.1 - 3 763
AK301873.1 - 4 599
BC002636.1 - 4 802 3606994
BC043232.1 - 6 1337 5295665
BI916125.1 - 477

View Full Product Details