Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001161527.1 | NP_001154999.1 | 6 | 904 | |
NM_001292016.1 | NP_001278945.1 | 5 | 941 | ||
NM_024309.3 | NP_077285.3 | 6 | 1190 | ||
GenBank mRNA | AF372839.1 | - | 6 | 1190 | |
AJ304866.1 | - | 6 | 1137 | ||
AK026176.1 | - | 6 | 903 | ||
AK096296.1 | - | 6 | 1181 | ||
AL137262.1 | - | 1 | 1724 | ||
BC002740.2 | - | 6 | 1148 | 3632736 | |
CB157550.1 | - | 2 | 155 |