Gene Transcripts

Gene Symbol
ZBED2
Entrez Gene ID
79413
Gene Name
zinc finger BED-type containing 2
Gene Aliases
Chromosome Location
Chr. 3: 111592900 - 111595335 on Build GRCh38
UniGene ID
Hs.136912
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024508.4 NP_078784.2 2 1649
GenBank mRNA BC003536.1 - 1 834
BC071956.1 - 1 1114 3928218

View Full Product Details