Gene Transcripts

Gene Symbol
TTC1
Entrez Gene ID
7265
Gene Name
tetratricopeptide repeat domain 1
Gene Aliases
TPR1
Chromosome Location
Chr. 5: 160009100 - 160065545 on Build GRCh38
UniGene ID
Hs.519718
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001282500.1 NP_001269429.1 3, 4 516
NM_003314.2 NP_003305.1 3, 4 519
GenBank mRNA AK222641.1 - 3, 4 470
AK222984.1 - 3, 4 464
AK315262.1 - 3, 4 474
BC000942.3 - 2, 3 424 3446109
DN989835.1 - 3, 4 472
U46570.1 - 3, 4 446

View Full Product Details