Gene Transcripts

Gene Symbol
PLXDC1
Entrez Gene ID
57125
Gene Name
plexin domain containing 1
Gene Aliases
TEM3, TEM7
Chromosome Location
Chr. 17: 39063303 - 39151649 on Build GRCh38
UniGene ID
Hs.125036
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020405.4 NP_065138.2 3 489
GenBank mRNA AF279144.2 - 3 359
AF378753.1 - 3 359
AK093589.1 - 3 510
AK127539.1 - 2 306
AK303699.1 - 2 176
AK312999.1 - 3 311
AY704670.1 - 3 359
AY704671.1 - 3 359
AY704672.1 - 3 305
BC036059.1 - 3 455 5298499
DA205327.1 - 3 489
KU257674.1 - 3 217

View Full Product Details