Gene Transcripts

Gene Symbol
THAP11
Entrez Gene ID
57215
Gene Name
THAP domain containing 11
Gene Aliases
CTG-B43a, CTG-B45d, HRIHFB2206, RONIN
Chromosome Location
Chr. 16: 67842310 - 67844195 on Build GRCh38
UniGene ID
Hs.632200
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020457.2 NP_065190.2 1 1769
GenBank mRNA BC012182.1 - 1 1728
L10378.1 - 1 1603
L10379.1 - 1 1603

View Full Product Details