Gene Transcripts

Gene Symbol
ZP4
Entrez Gene ID
57829
Gene Name
zona pellucida glycoprotein 4
Gene Aliases
ZBP, ZP1, ZPB, Zp-4
Chromosome Location
Chr. 1: 237877864 - 237890922 on Build GRCh38
UniGene ID
Hs.136241
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021186.4 NP_067009.1 11, 12 1786
GenBank mRNA AK314151.1 - 11, 12 1786
BC069521.1 - 11, 12 1499 7262334
DV080723.1 - 4, 5 449

View Full Product Details