Gene Transcripts

Gene Symbol
EVA1C
Entrez Gene ID
59271
Gene Name
eva-1 homolog C (C. elegans)
Gene Aliases
B18, B19, C21orf63, C21orf64, FAM176C, PRED34, SUE21
Chromosome Location
Chr. 21: 32411692 - 32515400 on Build GRCh38
UniGene ID
Hs.208358
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001286556.1 NP_001273485.1 8 1663
NM_001320744.1 NP_001307673.1 6 1407
NM_001320745.1 NP_001307674.1 8 1597
NM_058187.4 NP_478067.2 8 1672
NR_104472.1 7 1466
NR_135467.1 8 1771
XM_005261019.4 XP_005261076.1 6 1622
XM_006724038.3 XP_006724101.1 8 1790
XM_011529669.2 XP_011527971.1 8 1924
XM_017028418.1 XP_016883907.1 8 1790
XM_017028419.1 XP_016883908.1 7 1712
XM_017028420.1 XP_016883909.1 9 1713
XM_017028421.1 XP_016883910.1 8 1588
XM_017028422.1 XP_016883911.1 6 1463
XM_017028423.1 XP_016883912.1 5 1391
GenBank mRNA AF358258.1 - 8 1255
AK125013.1 - 7 2222
AY040087.1 - 7 1058
AY358787.1 - 8 1665
BC038710.1 - 8 1398 4813873

View Full Product Details