Gene Transcripts

Gene Symbol
TMEM2
Entrez Gene ID
23670
Gene Name
transmembrane protein 2
Gene Aliases
Chromosome Location
Chr. 9: 71683366 - 71769532 on Build GRCh38
UniGene ID
Hs.494146
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001135820.1 NP_001129292.1 4 1541
NM_013390.2 NP_037522.1 4 1541
XM_005251869.4 XP_005251926.1 4 1174
GenBank mRNA AB037833.2 - 4 1134
AF137030.1 - 4 1149
AK300405.1 - 4 1170
BC140773.1 - 4 1043 9021641
BC144510.1 - 4 1043
BC146780.1 - 4 1134 9007233
BC171782.1 - 4 1043

View Full Product Details