Gene Transcripts

Gene Symbol
PDCD4
Entrez Gene ID
27250
Gene Name
programmed cell death 4 (neoplastic transformation inhibitor)
Gene Aliases
H731
Chromosome Location
Chr. 10: 110871795 - 110900006 on Build GRCh38
UniGene ID
Hs.711490
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001199492.1 NP_001186421.1 12 2722
NM_014456.4 NP_055271.2 12 2764
NM_145341.3 NP_663314.1 13 2848
GenBank mRNA AL049932.1 - 1 997
BX647834.1 - 12 2604

View Full Product Details