Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_198182.2 | NP_937825.2 | 7 | 1163 | |
XM_005246159.3 | XP_005246216.1 | 6 | 1049 | ||
XM_006711882.2 | XP_006711945.1 | 7 | 1163 | ||
XM_006711884.3 | XP_006711947.1 | 6 | 1042 | ||
XM_011510343.2 | XP_011508645.1 | 7 | 1134 | ||
XM_017003900.1 | XP_016859389.1 | 7 | 1236 | ||
XM_017003901.1 | XP_016859390.1 | 7 | 1236 | ||
GenBank mRNA | AF198489.1 | - | 7 | 1101 | |
AF411210.1 | - | 3 | 518 | ||
AK293855.1 | - | 7 | 1210 | ||
AK296982.1 | - | 7 | 1241 | ||
AK312950.1 | - | 7 | 1133 | ||
BC067519.1 | - | 7 | 1044 | 6971723 | |
BC067520.1 | - | 7 | 1044 | 6971724 | |
BC067521.1 | - | 7 | 1044 | 6971725 |