Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001195139.1 | NP_001182068.1 | 10 | 1276 | |
NM_015386.2 | NP_056201.2 | 10 | 1276 | ||
XM_011522981.2 | XP_011521283.1 | 11 | 1436 | ||
GenBank mRNA | AB088369.1 | - | 11 | 1412 | |
AK022874.1 | - | 10 | 1256 | ||
AK026435.1 | - | 6 | 570 | ||
AK096557.1 | - | 8 | 994 | ||
AK293235.1 | - | 7 | 820 | ||
AK297557.1 | - | 11 | 1219 | ||
AL050101.1 | - | 6 | 578 | ||
BC006306.2 | - | 7 | 792 | ||
BC013347.2 | - | 6 | 584 | ||
BC027726.2 | - | 10 | 1250 | ||
BC072438.1 | - | 10 | 1256 | 6011802 | |
BC128144.1 | - | 10 | 1253 | ||
BC128145.1 | - | 10 | 1253 | ||
BX648010.1 | - | 8 | 1944 |