Gene Transcripts

Gene Symbol
CFAP61
Entrez Gene ID
26074
Gene Name
cilia and flagella associated protein 61
Gene Aliases
C20orf26, CaM-IP3, dJ1002M8.3, dJ1178H5.4
Chromosome Location
Chr. 20: 20052514 - 20360714 on Build GRCh38
UniGene ID
Hs.729168
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001167816.1 NP_001161288.1 3, 4 390
NM_015585.3 NP_056400.3 4, 5 496
XM_005260688.3 XP_005260745.1 4, 5 459
XM_005260689.2 XP_005260746.1 4, 5 459
XM_005260690.3 XP_005260747.1 4, 5 459
XM_005260691.3 XP_005260748.1 4, 5 459
XM_005260692.3 XP_005260749.1 4, 5 459
XM_005260693.3 XP_005260750.1 4, 5 459
XM_005260697.1 XP_005260754.1 4, 5 459
XM_011529211.1 XP_011527513.1 3, 4 317
XM_011529212.2 XP_011527514.1 4, 5 459
XM_011529213.1 XP_011527515.1 4, 5 459
XM_017027786.1 XP_016883275.1 4, 5 459
XM_017027787.1 XP_016883276.1 4, 5 459
XM_017027788.1 XP_016883277.1 4, 5 459
XM_017027789.1 XP_016883278.1 4, 5 459
XM_017027790.1 XP_016883279.1 4, 5 459
XM_017027795.1 XP_016883284.1 4, 5 459
GenBank mRNA AK304174.1 - 4, 5 459
AK307817.1 - 3, 4 317
AL117439.1 - 3, 4 328
BC028708.2 - 4, 5 458 4825941

View Full Product Details