Gene Transcripts
- Gene Symbol
- COQ4
- Entrez Gene ID
- 51117
- Gene Name
- coenzyme Q4
- Gene Aliases
- CGI-92, COQ10D7
- Chromosome Location
- Chr. 9: 128322486 - 128334072 on Build GRCh38
- UniGene ID
- Hs.98541
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AF151850.1 |
- |
5 |
783 |
|
View Full Product Details