Gene Transcripts

Gene Symbol
ANKFY1
Entrez Gene ID
51479
Gene Name
ankyrin repeat and FYVE domain containing 1
Gene Aliases
ANKHZN, BTBD23, ZFYVE14
Chromosome Location
Chr. 17: 4163371 - 4264026 on Build GRCh38
UniGene ID
Hs.696087
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001257999.1 NP_001244928.1 2, 3 446
NM_016376.3 NP_057460.3 2, 3 241
XM_005256679.3 XP_005256736.1 2, 3 288
XM_011523926.1 XP_011522228.1 2, 3 344
XM_011523928.2 XP_011522230.1 2, 3 326
XM_017024733.1 XP_016880222.1 2, 3 541
GenBank mRNA AB033081.2 - 2, 3 446
AB037360.1 - 2, 3 209
AK025483.1 - 2, 3 218
AK225170.1 - 2, 3 218
AK292930.1 - 2, 3 218
AK302712.1 - 2, 3 227
AL706226.1 - 2, 3 242
BC052308.1 - 2, 3 214
DA856905.1 - 2, 3 241

View Full Product Details