Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_003380.3 | NP_003371.2 | 5 | 1177 | |
XM_006717500.1 | XP_006717563.1 | 4 | 2252 | ||
GenBank mRNA | AF328728.1 | - | 4 | 893 | |
AK056766.1 | - | 6 | 846 | ||
AK091813.1 | - | 5 | 869 | ||
AK093924.1 | - | 3 | 848 | ||
AK097336.1 | - | 5 | 873 | ||
AK098444.1 | - | 5 | 747 | ||
AK222482.1 | - | 4 | 907 | ||
AK222507.1 | - | 4 | 909 | ||
AK222602.1 | - | 4 | 909 | ||
AK290643.1 | - | 4 | 789 | ||
BC000163.2 | - | 4 | 888 | 2985712 | |
BC030573.2 | - | 4 | 888 | 3637336 | |
BC066956.1 | - | 5 | 965 | 4823475 | |
CR407690.1 | - | 4 | 764 | ||
EU794672.1 | - | 4 | 909 | ||
KU178388.1 | - | 4 | 764 | ||
KU178389.1 | - | 5 | 381 | ||
M25246.1 | - | 4 | 268 | ||
X56134.1 | - | 4 | 807 | ||
Z19554.2 | - | 4 | 886 |