Gene Transcripts

Gene Symbol
PEX3
Entrez Gene ID
8504
Gene Name
peroxisomal biogenesis factor 3
Gene Aliases
PBD10A, TRG18
Chromosome Location
Chr. 6: 143450781 - 143490616 on Build GRCh38
UniGene ID
Hs.592832
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003630.2 NP_003621.1 10 1129
GenBank mRNA AB035307.1 - 10 1013
AJ001625.1 - 10 930
AJ131389.1 - 10 926
AK025644.1 - 995
AY277600.1 - 10 886
BC014551.1 - 10 1067 4333011
BC015506.1 - 10 1028 3914956
CR542062.1 - 10 867

View Full Product Details