Gene Transcripts

Gene Symbol
STK10
Entrez Gene ID
6793
Gene Name
serine/threonine kinase 10
Gene Aliases
LOK, PRO2729
Chromosome Location
Chr. 5: 172042070 - 172188342 on Build GRCh38
UniGene ID
Hs.744005
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005990.3 NP_005981.3 18 3041
XM_017009788.1 XP_016865277.1 17 2595
GenBank mRNA AB015718.1 - 18 2791
AK022960.1 - 10 1614
AK092814.1 - 1 247
AK307943.1 - 18 2574
AK313350.1 - 19 2796
AL133081.1 - 3 302
BC070077.1 - 18 2955 5267449

View Full Product Details