Gene Transcripts

Gene Symbol
STK10
Entrez Gene ID
6793
Gene Name
serine/threonine kinase 10
Gene Aliases
LOK, PRO2729
Chromosome Location
Chr. 5: 172042070 - 172188342 on Build GRCh38
UniGene ID
Hs.744005
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005990.3 NP_005981.3 19 3955
XM_017009788.1 XP_016865277.1 18 3509
GenBank mRNA AB015718.1 - 19 3705
AF119894.1 - 1 652
AK022960.1 - 11 2528
AK092814.1 - 2 1161
AL133081.1 - 4 1216
BC014413.1 - 1 624
BC070077.1 - 19 3869 5267449

View Full Product Details