Gene Transcripts

Gene Symbol
CDC16
Entrez Gene ID
8881
Gene Name
cell division cycle 16
Gene Aliases
ANAPC6, APC6, CDC16Hs, CUT9
Chromosome Location
Chr. 13: 114234845 - 114272675 on Build GRCh38
UniGene ID
Hs.374127
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AF164598.1 - 5 336
AK296424.1 - 5 523
BC017244.1 - 5 443 4877392
U18291.1 - 5 356

View Full Product Details