Gene Transcripts
- Gene Symbol
- OS9
- Entrez Gene ID
- 10956
- Gene Name
- OS9, endoplasmic reticulum lectin
- Gene Aliases
- ERLEC2, OS-9
- Chromosome Location
- Chr. 12: 57693955 - 57721557 on Build GRCh38
- UniGene ID
- Hs.527861
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001017956.2 |
NP_001017956.1
|
3, 4 |
602 |
|
NM_001017957.2 |
NP_001017957.1
|
3, 4 |
602 |
|
NM_001017958.2 |
NP_001017958.1
|
3, 4 |
602 |
|
NM_001261420.1 |
NP_001248349.1
|
3, 4 |
602 |
|
NM_001261421.1 |
NP_001248350.1
|
3, 4 |
602 |
|
NM_001261422.1 |
NP_001248351.1
|
3, 4 |
602 |
|
NM_001261423.1 |
NP_001248352.1
|
2, 3 |
425 |
|
NM_006812.3 |
NP_006803.1
|
3, 4 |
602 |
|
XM_005268581.1 |
XP_005268638.1
|
3, 4 |
534 |
|
XM_006719200.1 |
XP_006719263.1
|
3, 4 |
534 |
|
XM_006719201.1 |
XP_006719264.1
|
3, 4 |
534 |
|
GenBank mRNA |
AB002806.1 |
- |
3, 4 |
436 |
|
AK129781.1 |
- |
3, 4 |
414 |
|
AK130733.1 |
- |
3, 4 |
412 |
|
AK291374.1 |
- |
3, 4 |
484 |
|
AK293435.1 |
- |
2, 3 |
237 |
|
AK294870.1 |
- |
3, 4 |
414 |
|
AK296770.1 |
- |
3, 4 |
412 |
|
AK298532.1 |
- |
3, 4 |
412 |
|
AK303291.1 |
- |
3, 4 |
570 |
|
AK303858.1 |
- |
3, 4 |
412 |
|
AK304533.1 |
- |
3, 4 |
412 |
|
AL137691.1 |
- |
1, 2 |
340 |
|
AU132426.1 |
- |
3, 4 |
454 |
|
BC000532.2 |
- |
3, 4 |
407 |
2964645 |
BC007254.1 |
- |
3, 4 |
407 |
3027883 |
BC023513.2 |
- |
3, 4 |
397 |
4123782 |
CR456791.1 |
- |
3, 4 |
395 |
|
U41635.1 |
- |
3, 4 |
480 |
|
View Full Product Details