Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001195302.1 | NP_001182231.1 | 2 | 367 | |
NM_001204911.1 | NP_001191840.1 | 2 | 367 | ||
NM_024598.3 | NP_078874.2 | 2 | 367 | ||
XM_005256144.3 | XP_005256201.1 | 2 | 242 | ||
XM_011523328.2 | XP_011521630.1 | 2 | 343 | ||
XM_011523329.2 | XP_011521631.1 | 2 | 225 | ||
XM_011523330.1 | XP_011521632.1 | 2 | 333 | ||
GenBank mRNA | AK023216.1 | - | 2 | 339 | |
AK293220.1 | - | 2 | 177 | ||
AK298473.1 | - | 2 | 268 | ||
AK301494.1 | - | 2 | 317 | ||
AK303121.1 | - | 2 | 322 | ||
BC004415.1 | - | 2 | 303 | 3641317 | |
BC006291.2 | - | 2 | 293 | 4025993 | |
BC007774.2 | - | 2 | 297 | 4302289 | |
BC021554.1 | - | 2 | 287 | 4541229 | |
BM193105.1 | - | 2 | 340 | ||
DC363309.1 | - | 2 | 368 | ||
DC380401.1 | - | 2 | 367 |