Gene Transcripts

Gene Symbol
CNEP1R1
Entrez Gene ID
255919
Gene Name
CTD nuclear envelope phosphatase 1 regulatory subunit 1
Gene Aliases
C16orf69, NEP1-R1, NEP1R1, TMEM188, TMP125
Chromosome Location
Chr. 16: 50025206 - 50037088 on Build GRCh38
UniGene ID
Hs.59134
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001281789.1 NP_001268718.1 6 1099
NM_153261.5 NP_694993.2 7 1150
NR_104042.1 6 1076
NR_104043.1 5 1025
GenBank mRNA AK095420.1 - 6 1072
AK307132.1 - 6 1146
BC022550.1 - 5 976
BC036683.2 - 7 1079 5266084
CR933600.1 - 7 1095
FJ516381.1 - 6 1027

View Full Product Details