Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000965.4 | NP_000956.2 | 6 | 1320 | |
NM_001290216.2 | NP_001277145.1 | 9 | 1363 | ||
NM_001290217.1 | NP_001277146.1 | 6 | 778 | ||
NM_001290266.1 | NP_001277195.1 | 6 | 963 | ||
NM_001290276.1 | NP_001277205.1 | 6 | 1320 | ||
NM_001290300.1 | NP_001277229.1 | 6 | 784 | ||
NM_016152.3 | NP_057236.1 | 6 | 963 | ||
NR_110892.1 | 5 | 1159 | |||
GenBank mRNA | AB307688.1 | - | 6 | 851 | |
AF157483.1 | - | 6 | 962 | ||
AK292032.1 | - | 6 | 870 | ||
AK299756.1 | - | 6 | 938 | ||
AY501390.1 | - | 6 | 752 | ||
BC050415.2 | - | 4 | 546 | ||
BC060794.1 | - | 6 | 1002 | 30341884 | |
CD014021.1 | - | 5 | 706 | ||
CR536558.1 | - | 4 | 515 | ||
HQ692863.1 | - | 4 | 515 | ||
HQ692864.1 | - | 6 | 851 | ||
X07282.1 | - | 6 | 1194 | ||
Y00291.1 | - | 6 | 1172 |