Gene Transcripts

Gene Symbol
NEK2
Entrez Gene ID
4751
Gene Name
NIMA related kinase 2
Gene Aliases
HsPK21, NEK2A, NLK1, PPP1R111, RP67
Chromosome Location
Chr. 1: 211658256 - 211675630 on Build GRCh38
UniGene ID
Hs.153704
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001204182.1 NP_001191111.1 7 1226
NM_001204183.1 NP_001191112.1 7 1226
NM_002497.3 NP_002488.1 7 1226
XM_005273147.1 XP_005273204.1 7 1223
GenBank mRNA AK223353.1 - 7 1187
AK225522.1 - 7 1187
AK303092.1 - 8 1089
AY045701.1 - 7 1157
AY863109.1 - 3 430
BC043502.1 - 7 1214 5541026
BT019729.1 - 7 1075
BU517064.1 - 2 302
DQ786247.1 - 1 106
DQ786281.1 - 1 63
U11050.1 - 7 1209
Z29066.1 - 7 1157

View Full Product Details