Gene Transcripts

Gene Symbol
ESR2
Entrez Gene ID
2100
Gene Name
estrogen receptor 2
Gene Aliases
ER-BETA, ESR-BETA, ESRB, ESTRB, Erb, NR3A2
Chromosome Location
Chr. 14: 64226712 - 64338631 on Build GRCh38
UniGene ID
Hs.660607
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001040275.1 NP_001035365.1 2 804
NM_001214902.1 NP_001201831.1 1 426
NM_001271876.1 NP_001258805.1 1 426
NM_001271877.1 NP_001258806.1 1 426
NM_001291712.1 NP_001278641.1 7 2064
NM_001291723.1 NP_001278652.1 2 1079
NM_001437.2 NP_001428.1 2 804
NR_073496.1 2 1079
NR_073497.1 1 426
XM_017021079.1 XP_016876568.1 3 1468
XM_017021080.1 XP_016876569.1 3 2277
XM_017021081.1 XP_016876570.1 3 1373
XM_017021082.1 XP_016876571.1 3 1225
XM_017021083.1 XP_016876572.1 2 642
XM_017021084.1 XP_016876573.1 3 1468
GenBank mRNA AB006589.1 - 7 1611
AB006590.1 - 1 434
AB209620.1 - 1 432
AF051427.1 - 2 754
AF051428.1 - 2 754
AF060555.1 - 2 806
AF074599.1 - 1 164
AF124790.1 - 1 387
AK292370.1 - 2 641
AY438022.1 - 1 195
BC024181.2 - 2 621 4825937
DQ777076.1 - 1 336
DQ777077.1 - 1 336
DQ838582.1 - 1 336
DQ838583.1 - 1 336
HQ692820.1 - 1 336
HQ692821.1 - 1 336
KJ534833.1 - 1 336
X99101.1 - 1 195

View Full Product Details