Gene Transcripts
- Gene Symbol
- C5
- Entrez Gene ID
- 727
- Gene Name
- complement component 5
- Gene Aliases
- C5D, C5a, C5b, CPAMD4, ECLZB
- Chromosome Location
- Chr. 9: 120952335 - 121075174 on Build GRCh38
- UniGene ID
- Hs.494997
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001317163.1 |
NP_001304092.1
|
10, 11 |
1480 |
|
NM_001317164.1 |
NP_001304093.1
|
10, 11 |
1151 |
|
NM_001735.2 |
NP_001726.2
|
10, 11 |
1154 |
|
XM_011518980.2 |
XP_011517282.1
|
11, 12 |
1523 |
|
XM_017015102.1 |
XP_016870591.1
|
12, 13 |
1675 |
|
XM_017015103.1 |
XP_016870592.1
|
10, 11 |
1308 |
|
XM_017015104.1 |
XP_016870593.1
|
12, 13 |
1675 |
|
GenBank mRNA |
AB209031.1 |
- |
10, 11 |
1480 |
|
AK310774.1 |
- |
10, 11 |
1151 |
|
AK310780.1 |
- |
10, 11 |
1153 |
|
BC022299.1 |
- |
10, 11 |
1155 |
|
BC113738.1 |
- |
10, 11 |
1149 |
8322790 |
BC113740.1 |
- |
10, 11 |
1149 |
8322792 |
M57729.1 |
- |
10, 11 |
1136 |
|
View Full Product Details