Gene Transcripts

Gene Symbol
FCHSD2
Entrez Gene ID
9873
Gene Name
FCH and double SH3 domains 2
Gene Aliases
NWK, SH3MD3
Chromosome Location
Chr. 11: 72836743 - 73142682 on Build GRCh38
UniGene ID
Hs.744959
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014824.2 NP_055639.2 17 2003
XM_011545409.1 XP_011543711.1 17 1885
XM_011545410.2 XP_011543712.1 17 1818
XM_017018632.1 XP_016874121.1 17 2062
GenBank mRNA AB018312.1 - 16 1853
AK296376.1 - 17 1745
AK297928.1 - 17 1887
AL133567.1 - 12 1371
BC017751.1 - 5 553

View Full Product Details