Gene Transcripts

Gene Symbol
ATP13A4
Entrez Gene ID
84239
Gene Name
ATPase 13A4
Gene Aliases
Chromosome Location
Chr. 3: 193398967 - 193581725 on Build GRCh38
UniGene ID
Hs.674423
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032279.3 NP_115655.2 5, 6 867
XM_011513232.2 XP_011511534.1 5, 6 614
XM_017007318.1 XP_016862807.1 5, 6 503
XM_017007319.1 XP_016862808.1 5, 6 614
GenBank mRNA AK095277.1 - 5, 6 652
AY823162.1 - 5, 6 544
BC101496.1 - 5, 6 638 8069002

View Full Product Details