Gene Transcripts

Gene Symbol
WDR36
Entrez Gene ID
134430
Gene Name
WD repeat domain 36
Gene Aliases
GLC1G, TA-WDRP, TAWDRP, UTP21
Chromosome Location
Chr. 5: 111092172 - 111130502 on Build GRCh38
UniGene ID
Hs.533237
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_139281.2 NP_644810.1 12 1521
GenBank mRNA AF385437.1 - 12 1521
AK074202.1 - 6 571
AK095312.1 - 12 1379
AL832494.1 - 11 1323
BC133025.1 - 12 1481 40147047
BC136517.1 - 12 1481 9020507
CR749211.1 - 7 721

View Full Product Details