Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001134658.1 | NP_001128130.1 | 3 | 727 | |
NM_153226.2 | NP_694958.1 | 3 | 724 | ||
XM_011539371.2 | XP_011537673.1 | 3 | 692 | ||
GenBank mRNA | AK091309.1 | - | 3 | 724 | |
BC041432.1 | - | 2 | 456 | ||
BC104814.1 | - | 3 | 698 | 8143817 | |
BC143283.1 | - | 3 | 647 | 9051786 | |
BC143284.1 | - | 3 | 698 | 9051787 |