Gene Transcripts

Gene Symbol
FIBIN
Entrez Gene ID
387758
Gene Name
fin bud initiation factor homolog (zebrafish)
Gene Aliases
Chromosome Location
Chr. 11: 26994081 - 26997087 on Build GRCh38
UniGene ID
Hs.712718
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_203371.1 NP_976249.1 1 1408
GenBank mRNA AK075535.1 - 1 970
BC026873.1 - 1 1179 4938507

View Full Product Details