Gene Transcripts

Gene Symbol
MAMDC4
Entrez Gene ID
158056
Gene Name
MAM domain containing 4
Gene Aliases
AEGP
Chromosome Location
Chr. 9: 136852367 - 136860799 on Build GRCh38
UniGene ID
Hs.376780
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_206920.2 NP_996803.2 7 826
GenBank mRNA AL137659.1 - 7 1212
AY358419.1 - 7 826

View Full Product Details