Gene Transcripts

Gene Symbol
SEPT14
Entrez Gene ID
346288
Gene Name
septin 14
Gene Aliases
Chromosome Location
Chr. 7: 55793544 - 55862789 on Build GRCh38
UniGene ID
Hs.453629
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_207366.2 NP_997249.2 10 1940
XM_011515373.2 XP_011513675.1 10 1924
GenBank mRNA AK126048.1 - 5 1228

View Full Product Details