Gene Transcripts

Gene Symbol
C8orf86
Entrez Gene ID
389649
Gene Name
chromosome 8 open reading frame 86
Gene Aliases
Chromosome Location
Chr. 8: 38510834 - 38528662 on Build GRCh38
UniGene ID
Hs.546586
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001303531.1 NP_001290460.1 2 847
NM_207412.2 NP_997295.1 3 1020
GenBank mRNA AK125570.1 - 3 1020
BC137511.1 - 3 1015 9021511
BC137512.1 - 3 1015 9021512
BC139744.1 - 2 842 40107968

View Full Product Details