Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_008851.4 | NP_032877.1 | 23 | 4396 | |
GenBank mRNA | AF006467.1 | - | 24 | 3971 | |
AK155377.1 | - | 23 | 4869 | ||
AK209641.1 | - | 1 | 233 | ||
AK217085.1 | - | 1 | 233 | ||
AK218358.1 | - | 1 | 233 | ||
BC044893.1 | - | 23 | 3901 | 4481550 | |
BC048150.1 | - | 23 | 4050 | 5706382 | |
X98655.1 | - | 1 | 289 | ||
Y08922.1 | - | 23 | 4397 |