Gene Transcripts

Gene Symbol
Pon1
Entrez Gene ID
18979
Gene Name
paraoxonase 1
Gene Aliases
Pon
Chromosome Location
Chr. 6: 5168090 - 5193987 on Build GRCm38
UniGene ID
Mm.237657
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011134.3 NP_035264.2 4 544
GenBank mRNA BC012706.1 - 4 417
U32684.1 - 4 426
U72636.1 - 4 426

View Full Product Details