Gene Transcripts

Gene Symbol
Mios
Entrez Gene ID
252875
Gene Name
missing oocyte, meiosis regulator, homolog (Drosophila)
Gene Aliases
C81488
Chromosome Location
Chr. 6: 8209004 - 8236274 on Build GRCm38
UniGene ID
Mm.257151
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145374.2 NP_663349.2 3 1290
GenBank mRNA BC020002.1 - 3 1288
BC025652.1 - 1 358
CX565910.1 - 1 478

View Full Product Details