Gene Transcripts

Gene Symbol
Camkv
Entrez Gene ID
235604
Gene Name
CaM kinase-like vesicle-associated
Gene Aliases
1G5, BB074618, BC017634
Chromosome Location
Chr. 9: 107935851 - 107949691 on Build GRCm38
UniGene ID
Mm.274540
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145621.2 NP_663596.1 6 653
GenBank mRNA AK147323.1 - 6 641
BC017634.1 - 6 653 4527857
BC022169.1 - 5 862
BC086762.1 - 5 851
BC111033.1 - 6 648 5368225

View Full Product Details