Gene Transcripts
- Gene Symbol
- FAM189A2
- Entrez Gene ID
- 9413
- Gene Name
- family with sequence similarity 189 member A2
- Gene Aliases
- C9orf61, X123
- Chromosome Location
- Chr. 9: 69324572 - 69392461 on Build GRCh38
- UniGene ID
- Hs.118003
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001127608.1 |
NP_001121080.1
|
7, 8 |
730 |
|
NM_004816.3 |
NP_004807.3
|
7, 8 |
1046 |
|
XM_005252307.4 |
XP_005252364.1
|
7, 8 |
1607 |
|
XM_017015324.1 |
XP_016870813.1
|
6, 7 |
791 |
|
XM_017015325.1 |
XP_016870814.1
|
7, 8 |
730 |
|
GenBank mRNA |
AK056152.1 |
- |
5, 6 |
674 |
|
AK126127.1 |
- |
7, 8 |
1046 |
|
BC021685.1 |
- |
4, 5 |
459 |
|
BC113683.1 |
- |
7, 8 |
689 |
8322735 |
BC113685.1 |
- |
7, 8 |
689 |
8322737 |
BX641153.1 |
- |
4, 5 |
5899 |
|
View Full Product Details