Gene Transcripts

Gene Symbol
Nkain3
Entrez Gene ID
269513
Gene Name
Na+/K+ transporting ATPase interacting 3
Gene Aliases
C530048M05, E130310K16Rik
Chromosome Location
Chr. 4: 20118336 - 20780310 on Build GRCm38
UniGene ID
Mm.317473
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001290410.1 NP_001277339.1 4 770
NM_172987.2 NP_766575.2 4 770
GenBank mRNA AK049776.1 - 4 970
AK053823.1 - 4 655
AK141456.1 - 4 670
BC075683.1 - 4 770 30613986
EF058052.1 - 4 417
EF058053.1 - 4 417
EF058054.1 - 4 417

View Full Product Details